Learn More
Fetuin Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-99828JF646
Product Code. 18462489
  • € 407.00 / 0.10mL
Description
Fetuin Polyclonal antibody specifically detects Fetuin in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rabbit
Produced in rabbits immunized with purified, recombinant Human Fetuin (Accession#: NP_055190.2; Met 1-Pro 382)