Learn More
Fetuin Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-99828PCP
Product Code. 18697647
  • € 407.00 / 0.10mL
Description
Fetuin Polyclonal antibody specifically detects Fetuin in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ELISA
fetuin B, fetuin-B, Fetuin-like protein IRL685,16G2, Gugu, IRL685
26998
Store at 4C in the dark.