Learn More
Fexinidazole, MedChemExpress
Product Code. 16458517
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16458517
Brand: Medchem Express HY13801
MedChemExpress Fexinidazole (HOE 239) is an orally active, potent nitroimidazole antitrypanosomal drug. Fexinidazole shows trypanocidal activity against T. brucei subspecies and strains with IC50s of 0.7-3.3 μM (0.2-0.9 μg/ml). Fexinidazol has the potential for human sleeping sickness (HAT) caused by infection with T. brucei.
Fexinidazole (HOE 239) is an orally active, potent nitroimidazole antitrypanosomal drug. Fexinidazole shows trypanocidal activity against T. brucei subspecies and strains with IC50s of 0.7-3.3 μM (0.2-0.9 μg/ml). Fexinidazol has the potential for human sleeping sickness (HAT) caused by infection with T. brucei.
Specifications
Chemical Name or Material Fexinidazole
Physical Form Solid
CAS 59729-37-2
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C12H13N3O3S
For Use With (Application) COVID-19-immunoregulation
Synonym HOE 239
Solubility Information DMSO : 50 mg/mL (179.01 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=[N+](C1=CN=C(COC2=CC=C(SC)C=C2)N1C)[O-]
Molecular Weight (g/mol) 279.31
Formula Weight 279.31
Percent Purity 98.65%
Grade Research
Show More Show Less
Research purposes only