missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FEZ1 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76395C
This item is not returnable.
View return policy
Description
FEZ1 Polyclonal antibody specifically detects FEZ1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| FEZ1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| DyLight 650 | |
| fasciculation and elongation protein zeta 1 (zygin I), fasciculation and elongation protein zeta-1, Zygin I, zygin-1 | |
| Antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human FEZ1. The immunogen is located within amino acids 270 - 320 of FEZ1. Amino Acid Squence: KQKEQRELMKKRRKEKGL | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 9638 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction