missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FEZ1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16537821
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16537821 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16537821 Supplier Invitrogen™ Supplier No. PA590412

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MEAPLVSLDE EFEDLRPSCS EDPEEKPQCF YGSSPHHLED PSLSELENFS SEIISFKSME DLVNEFDEKL NVCFRNYNAK TENLAPVKNQ LQIQEEEETL QDEEVWDALT DNYIPSLSED WRDPNIEALN GNCSDTEIHE KEEEEFNEKS ENDSGINEEP; Positive Samples: Mouse brain, Rat brain; Cellular Location: Cell membrane, Cytoplasm, centrosome, cytoskeleton, microtubule organizing center.

This gene is an ortholog of the C. elegans unc-76 gene, which is necessary for normal axonal bundling and elongation within axon bundles. Expression of this gene in C. elegans unc-76 mutants can restore to the mutants partial locomotion and axonal fasciculation, suggesting that it also functions in axonal outgrowth. The N-terminal half of the gene product is highly acidic. Alternatively spliced transcript variants encoding different isoforms of this protein have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FEZ1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Fez1
Gene Accession No. P97577, Q8K0X8
Gene Alias F37; fasciculation and elongation protein zeta 1; fasciculation and elongation protein zeta 1 (zygin I); fasciculation and elongation protein zeta-1; Fez1; protein kinase C-binding protein Zeta1; UNC76; UNC-76; zygin 1; zygin I; Zygin-1
Gene Symbols Fez1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human FEZ1 (NP_0050941).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235180, 81730
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.