Learn More
Tocris Bioscience™ FFN 102 mesylate
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
FFN 102 mesylate is a pH responsive fluorescent false neurotransmitter (FFN). Selective dopamine transporter (DAT) and VMAT2 substrate. Exhibits no significant binding to a panel of 38 CNS receptors, including dopamine and serotonin receptors. Inhibits dopamine uptake. Excitation maxima are 340 nm at pH 5 and 370 nm at pH 7.5. Emission maximum is 435 nm at both pH 5 and 7.5.
Specifications
Specifications
| CAS | 1883548-92-2 |
| Quantity | 10 mg |
| Product Type | FFN 102 mesylate |
| Purity | >99% |
| Molecular Weight (g/mol) | 335.76 |
| Inhibitors | FFN 102 mesylate |