missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-10 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99399R
This item is not returnable.
View return policy
Description
FGF-10 Polyclonal antibody specifically detects FGF-10 in Human samples. It is validated for Western Blot
Specifications
| FGF-10 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| FGF-10, fibroblast growth factor 10, Keratinocyte growth factor 2, produced by fibroblasts of urinary bladder lamina propria | |
| Produced in rabbits immunized with purified, recombinant Human FGF-10 (Accession#: NP_004456.1; Gln38-Ser208) | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication | |
| 2255 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction