missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-13 Antibody (S235-22), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42178AF405
This item is not returnable.
View return policy
Description
FGF-13 Monoclonal specifically detects FGF-13 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| FGF-13 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FGF13 | |
| Synthetic peptide amino acids 2-18 (AAAIASSLIRQKRQARE) of human FHF2A. 100% identical to rat, 94% identical to mouse. >80% identity with FGF12A/FHF1A, FGF14A/FHF4A and FGF11A/FHF3A. | |
| 0.1 mL | |
| Primary | |
| Greater then 80% identity with FGF12A/FHF1A, FGF14A/FHF4A and FGF11A/FHF3A. Detects approx 30kDa. Does not cross-react with FGF13B/FHF2B. Cross reacts with FGF12A/FHF1A and FGF14A/FHF4A. | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S235-22 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13 | |
| Mouse | |
| Protein G purified | |
| Angiogenesis, Biologically Active Proteins, Neuroscience | |
| 2258 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction