missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-16 Antibody (MM0283-6G2), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12297DL594
This item is not returnable.
View return policy
Description
FGF-16 Monoclonal antibody specifically detects FGF-16 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| FGF-16 | |
| Monoclonal | |
| DyLight 594 | |
| FGF-16, fibroblast growth factor 16 | |
| Human recombinant FGF-16 | |
| 0.1 mL | |
| Angiogenesis | |
| 8823 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0283-6G2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction