missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-19 Antibody (032), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90078V
This item is not returnable.
View return policy
Description
FGF-19 Monoclonal specifically detects FGF-19 in Human samples. It is validated for ELISA.
Specifications
| FGF-19 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Human FGF6 Human FGF9 Human FGF10 Human FGF17 Human FGF18 Human FGF acidic Human FGF basic | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 32 | |
| ELISA | |
| FGF-19, fibroblast growth factor 19 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human FGF-19 (O95750; Phe27-Lys216). | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Neuroscience | |
| 9965 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction