missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-21 Antibody (OTI2F10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-00645
This item is not returnable.
View return policy
Description
FGF-21 Monoclonal specifically detects FGF-21 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| FGF-21 | |
| Monoclonal | |
| 1.4 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:150 | |
| FGF21 | |
| Human recombinant protein fragment corresponding to amino acids 29-209 of human FGF21(NP_061986) produced in E.coli. | |
| 0.1 mL | |
| Angiogenesis, Cancer | |
| 26291 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI2F10 | |
| Unconjugated | |
| FGF-21, fibroblast growth factor 21 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction