missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-22 Antibody (MM0286-4F28), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12300B
This item is not returnable.
View return policy
Description
FGF-22 Monoclonal specifically detects FGF-22 in Human samples. It is validated for Western Blot.
Specifications
| FGF-22 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| FGF22 | |
| Human recombinant FGF-22 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MM0286-4F28 | |
| Western Blot | |
| FGF-22, fibroblast growth factor 22 | |
| Mouse | |
| Protein G purified | |
| Angiogenesis | |
| 27006 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction