missing translation for 'onlineSavingsMsg'
Learn More

FGF-23 Antibody (FGF23/132), Novus Biologicals™

Mouse Monoclonal Antibody

€ 275.00 - € 518.00

Specifications

Antigen FGF-23
Clone FGF23/132
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18682124
View Documents
Novus Biologicals
NBP3-20358-20ug
20 μg
€ 275.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18631564
View Documents
Novus Biologicals
NBP3-20358-100ug
100 μg
€ 518.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FGF-23 Monoclonal antibody specifically detects FGF-23 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

FGF-23
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Angiogenesis, Biologically Active Proteins, Cancer
10mM PBS with 0.05% BSA
8074
IgG
Protein A or G purified
FGF23/132
Immunohistochemistry-Paraffin 1-2 μg/mL
Monoclonal
Purified
RUO
Human
ADHR, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor
Recombinant full-length human FGF-23 protein
Primary
Store at 4&drg;C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.