missing translation for 'onlineSavingsMsg'
Learn More

FGF-23 Antibody (FGF23/4168), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-08383AF405

Product Code. 18544134

  • € 608.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FGF-23 Monoclonal antibody specifically detects FGF-23 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FGF-23
Monoclonal
Alexa Fluor 405
ADHR, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor
Recombinant fragment (around aa25-251) of human FGF-23 protein (exact sequence is proprietary) (Uniprot: Q9GZV9)
100 μg
Angiogenesis, Biologically Active Proteins, Cancer
8074
Store at 4°C in the dark.
IgG2c κ
Immunohistochemistry (Paraffin), Protein Array
FGF23/4168
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.