missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-23 Antibody (FGF23/4168), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08383CL1
This item is not returnable.
View return policy
Description
FGF-23 Monoclonal antibody specifically detects FGF-23 in Human samples. It is validated for Immunohistochemistry (Paraffin),Protein Array
Specifications
| FGF-23 | |
| Monoclonal | |
| CoraFluor 1 | |
| ADHR, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor | |
| Recombinant fragment (around aa25-251) of human FGF-23 protein (exact sequence is proprietary) (Uniprot: Q9GZV9) | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Cancer | |
| 8074 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2c κ |
| Immunohistochemistry (Paraffin), Protein Array | |
| FGF23/4168 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu