missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-4 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76865DL594
This item is not returnable.
View return policy
Description
FGF-4 Polyclonal antibody specifically detects FGF-4 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| FGF-4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 594 | |
| fibroblast growth factor 4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST | |
| Antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of the human FGF4. The immunogen is located within the last 50 amino acids of FGF4. Amino Acid Squence: SKNGKTKKGNRVSPTMK | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Cancer | |
| 2249 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction