missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-8 Rabbit anti-Human, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90327-100UL
This item is not returnable.
View return policy
Description
FGF-8 Monoclonal specifically detects FGF-8 in Human samples. It is validated for ELISA.
Specifications
| FGF-8 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 1 | |
| ELISA 1:5000-1:10000 | |
| AIGFKAL6, Androgen-induced growth factor, FGF-8, fibroblast growth factor 8, fibroblast growth factor 8 (androgen-induced), HBGF-8, Heparin-binding growth factor 8, MGC149376 | |
| Recombinant Human FGF-8 Protein | |
| 100 μg | |
| Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication | |
| 2253 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction