missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-9 Antibody (MM0292-4D25), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12306AF750
This item is not returnable.
View return policy
Description
FGF-9 Monoclonal specifically detects FGF-9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| FGF-9 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| FGF9 | |
| Human recombinant FGF-9 | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0292-4D25 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FGF-9, Fibroblast growth factor 9, fibroblast growth factor 9 (glia-activating factor), GAF, glia-activating factor, HBFG-9, HBGF-9, Heparin-binding growth factor 9, MGC119914, MGC119915, SYNS3 | |
| Mouse | |
| 0.1 mL | |
| 2254 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction