missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF acidic/FGF1 Antibody (V8P1G5*F2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50406F
This item is not returnable.
View return policy
Description
FGF acidic/FGF1 Monoclonal specifically detects FGF acidic/FGF1 in Zebrafish samples. It is validated for ELISA, Immunohistochemistry.
Specifications
| FGF acidic/FGF1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| FGF1 | |
| Ovalbumin-conjugated synthetic peptide LPRQVEQEED (C-terminal sequence) | |
| 0.1 mL | |
| Angiogenesis, Cancer, Stem Cell Markers | |
| 2246 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| ELISA, Immunohistochemistry | |
| V8P1G5*F2 | |
| ELISA, Immunohistochemistry | |
| AFGF, alpha, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction