missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF basic/FGF2/bFGF Antibody (101), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89204AF532
This item is not returnable.
View return policy
Description
FGF basic/FGF2/bFGF Monoclonal antibody specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for ELISA
Specifications
| FGF basic/FGF2/bFGF | |
| Monoclonal | |
| Alexa Fluor 532 | |
| Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human FGF basic/FGF2/bFGF (Accession#: NP_001997.5; Pro 143-Ser 288). | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Cancer | |
| 2247 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 101 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction