missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-BP Antibody (OTI2B6), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71527F
This item is not returnable.
View return policy
Description
FGF-BP Monoclonal specifically detects FGF-BP in Human samples. It is validated for Immunohistochemistry.
Specifications
| FGF-BP | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| FGFBP1 | |
| Recombinant protein fragment corresponding to amino acids 24-234 of human FGF-BP (NP_005121) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Immunohistochemistry | |
| OTI2B6 | |
| Immunohistochemistry | |
| 17 kDa heparin-binding growth factor-binding protein, FGF-BP, FGF-BP1, FGFBP-1, FGFBPFGF-binding protein 1, fibroblast growth factor binding protein 1, fibroblast growth factor-binding protein 1, HBp17, HBP1717 kDa HBGF-binding protein, heparin-binding growth factor binding protein | |
| Mouse | |
| Affinity Purified | |
| Angiogenesis, Cell Cycle and Replication | |
| 9982.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction