Learn More
Invitrogen™ FGF11 Polyclonal Antibody
GREENER_CHOICE
Product Code. 30535134
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30535134 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30535134 Supplier Invitrogen™ Supplier No. PA5148515
Sheep Polyclonal Antibody
Reconstitute at 0.2 mg/mL in sterile PBS.
Probably involved in nervous system development and function.
TRUSTED_SUSTAINABILITY
Specifications
Antigen FGF11
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene FGF11
Gene Accession No. Q92914
Gene Alias FGF; Fgf11; Fgf-11; Fhf3; FHF-3; Fibroblast growth factor; Fibroblast growth factor 11; fibroblast growth factor homologous factor 3
Gene Symbols FGF11
Host Species Sheep
Immunogen E. coli-derived recombinant human FGF-11.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2256
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG