Learn More
Invitrogen™ FGF13 Monoclonal Antibody (N235/22), PerCP

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545653
Description
100% identical to rat, 94% identical to mouse. >80% identity with FGF12A/FHF1A, FGF14A/FHF4A and FGF11A/FHF3A. 1 μg/mL of MA5-45653 was sufficient for detection of FGFA/FHFA (pan) in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 30kDa. Does not cross-react with FGF13B/FHF2B. Cross reacts with FGF12A/FHF1A and FGF14A/FHF4A. This antibody was formerly sold as clone S235-22.
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. This gene is located in a region on chromosome X, which is associated with Borjeson-Forssman-Lehmann syndrome (BFLS), making it a possible candidate gene for familial cases of the BFLS, and for other syndromal and nonspecific forms of X-linked mental retardation mapping to this region. Alternative splicing of this gene at the 5' end results in several transcript variants encoding different isoforms with different N-termini.
Specifications
| FGF13 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| P70377, Q92913, Q9ERW3 | |
| FGF13 | |
| Synthetic peptide amino acids 2-18 (AAAIASSLIRQKRQARE) of human FHF2A. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| N235/22 | |
| PerCP | |
| FGF13 | |
| AFGF; ECGF; ECGFA; ECGFB; FGF; FGF 1; FGF alpha; FGF13; FGF-13; FGF13A; FGF2; FGFA; Fhf2; FHF-2; Fibroblast growth factor; fibroblast growth factor 13; Fibroblast growth factor homologous factor 2; GLIO703; HBGF 1; HBGF1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 14168, 2258, 84488 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.