missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF14 Rabbit anti-Human, Clone: 107, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90235-50UL
This item is not returnable.
View return policy
Description
FGF14 Monoclonal specifically detects FGF14 in Human samples. It is validated for ELISA.
Specifications
| FGF14 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 107 | |
| ELISA 1:5000-1:10000 | |
| bA397O8.2, FGF-14, FHF4FHF-4, fibroblast growth factor 14, Fibroblast growth factor homologous factor 4, SCA27MGC119129 | |
| Recombinant Human FGF14 protein | |
| 50 μg | |
| Angiogenesis, Neuroscience | |
| 2259 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction