missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF18 Antibody (PSH01-24), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32338
This item is not returnable.
View return policy
Description
FGF18 Monoclonal antibody specifically detects FGF18 in Human,Mouse samples. It is validated for Western Blot
Specifications
| FGF18 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| FGF-18ZFGF5, fibroblast growth factor 18, zFGF5 | |
| Recombinant protein within mouse FGF18 aa 1-207/ 207. (Uniprot: O76093) | |
| 100 μL | |
| Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication | |
| 8817 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| PSH01-24 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction