Learn More
Invitrogen™ FGF19 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100905
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FGF19.
FGF19, Fibroblast growth factor 19, is a protein that in humans is encoded by the FGF19 gene. The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. The FGF19 gene is mapped to 11q13.3. The deduced 216-amino acid FGF19 protein contains asignal sequence and 2 cysteine residues that are conserved in the FGF family. Expression of this gene was detected only in fetal but not adult brain tissue. Synergistic interaction of the chick homolog and Wnt-8c has been shown to be required for initiation of inner ear development. FGF19 stimulates hepatic protein and glycogen synthesis but does not induce lipogenesis. The effects of FGF19 are independent of the activity of either insulin or the protein kinase Akt and, instead, are mediated through a mitogen-activated protein kinase signaling pathway that activates components of the protein translation machinery and stimulates glycogen synthase activity. The orthologous protein in mouse is FGF15, which shares about 50% amino acid identity and has similar functions. Together they are often referred to as FGF15/19.
Specifications
| FGF19 | |
| Polyclonal | |
| Unconjugated | |
| FGF19 | |
| FGF; Fgf15; FGF19; FGF-19; Fibroblast growth factor; fibroblast growth factor 15; fibroblast growth factor 19; UNQ334/PRO533 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 170582, 9965 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95750 | |
| FGF19 | |
| A synthesized peptide derived from human FGF19(Accession O95750), corresponding to amino acid residues M168-K216. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |