missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGF20 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148602
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
FGF-20 is a secreted, heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-20 signals through FGFR 2c and 3c, and is expressed during limb and brain development.
Specifications
| FGF20 | |
| Polyclonal | |
| Unconjugated | |
| FGF20 | |
| FGF; Fgf20; FGF-20; Fibroblast growth factor; Fibroblast growth factor 20; RHDA2 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 26281 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9NP95 | |
| FGF20 | |
| E. coli-derived recombinant human FGF-20, Pro3-Thr211. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction