Learn More
FGF21 Monoclonal Antibody (OTI1H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA502816
Encompass_Preferred
 View more versions of this product
Product Code. 17557831
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The function of this growth factor has not yet been determined.
Specifications
Specifications
FGF21
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NSA1
FGF21
Human recombit protein fragment corresponding to amino acids 29-209 of human FGF21 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Western Blot
OTI1H5
Unconjugated
FGF21
fibroblast growth factor 21
Mouse
Affinity Chromatography
RUO
26291
-20° C, Avoid Freeze/Thaw Cycles