Learn More
Invitrogen™ FGF23 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120972
Description
Positive test controls include: Mouse brain, Rat brain. The target is usually found in the following locations: Secreted. Immunogen sequence: YPNASPLLGS SWGGLIHLYT ATARNSYHLQ IHKNGHVDGA PHQTIYSALM IRSEDAGFVV ITGVMSRRYL CMDFRGNIFG SHYFDPENCR FQHQTLENGY DVYHSPQYHF LVSLGRAKRA FLPGMNPPPY SQFLSRRNEI PLIHFNTPIP RRHTRSAEDD SERDPLNVLK PRARMTPAPA SCSQELPSAE DNSPMASDPL GVVRGGRVNT HAGGTGPEGC RPFAKFI.
The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The product of this gene inhibits renal tubular phosphate transport. This gene was identified by its mutations associated with autosomal dominant hypophosphatemic rickets, an inherited phosphate wasting disorder. Abnormally high level expression of this gene was found in oncogenic hypophosphatemic osteomalacia, a phenotypically similar disease caused by abnormal phosphate metabolism. Mutations in this gene have also been shown to cause familial tumoral calcinosis with hyperphosphatemia.
Specifications
| FGF23 | |
| Polyclonal | |
| Unconjugated | |
| FGF23 | |
| ADHR; FGF; Fgf23; FGF-23; FGFN; Fibroblast growth factor; Fibroblast growth factor 23; Fibroblast growth factor 23 C-terminal peptide; Fibroblast growth factor 23 N-terminal peptide; HPDR2; HYPF; Phosphatonin; PHPTC; tumor-derived hypophosphatemia inducing factor; Tumor-derived hypophosphatemia-inducing factor; UNQ3027/PRO9828 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 170583, 64654, 8074 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.84 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q8VI82, Q9EPC2, Q9GZV9 | |
| FGF23 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.