missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGF4 Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHFGF4X10
Additional Details : Weight : 2.50000kg
Description
Fibroblast growth factor 4 (FGF4) is a member of the fibroblast growth factor (FGF) family that possess broad mitogenic and cell survival activities and play key roles in growth and survival of stem cells during embryogenesis, tissue regeneration, and carcinogenesis. FGF4 was identified by its strong oncogenic transforming activity and is a potent angiogenic factor, expressed in several highly vascularized tumors and also in adult mouse testis, intestine, and brain. Studies on the mouse homolog suggests a function in bone morphogenesis and limb development through the sonic hedgehog (SHH) signaling pathway. Furthermore, FGF4 regulates neural progenitor cell proliferation and neuronal differentiation. Recent studies show a growth-promoting role for FGF4 in human embryonic stem cells and a putative feedback inhibition mechanism by a novel FGF4 splice isoform that may serve to promote differentiation at a later stages of development.Specifications
P08620 | |
50 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Fgf4 | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
24.69 to 18,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
2249 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 50 μL|Serum, 50 μL|Supernatant, 100 μL | |
FGF4 | |
4 hr 45 min |