missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGF9 Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHFGF9X10
Additional Details : Weight : 2.50000kg
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein was isolated as a secreted factor that exhibits a growth-stimulating effect on cultured glial cells. In nervous system, this protein is produced mainly by neurons and may be important for glial cell development. Expression of the mouse homolog of this gene was found to be dependent on Sonic hedgehog (Shh) signaling. Mice lacking the homolog gene displayed a male-to-female sex reversal phenotype, which suggested a role in testicular embryogenesis.Specifications
P31371 | |
200 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
FGF9 | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
358.4 to 35,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
2254 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 50 μL|Serum, 50 μL|Supernatant, 100 μL | |
FGF9 | |
4 hr 45 min |