missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFBP1 Monoclonal Antibody (OTI2B6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808309
This item is not returnable.
View return policy
Description
FGFBP1 encodes a secreted fibroblast growth factor carrier protein. The encoded protein plays a critical role in cell proliferation, differentiation and migration by binding to fibroblast growth factors and potentiating their biological effects on target cells. The encoded protein may also play a role in tumor growth as an angiogenic switch molecule, and expression of this gene has been associated with several types of cancer including pancreatic and colorectal adenocarcinoma. A pseudogene of this gene is also located on the short arm of chromosome 4.Specifications
| FGFBP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q14512 | |
| FGFBP1 | |
| Human recombit protein fragment corresponding to amino acids 24-234 of human FGFBP1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2B6 | |
| Unconjugated | |
| FGFBP1 | |
| FGF-BP, FGF-BP1, FGFBP, FGFBP-1, HBP17 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9982 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction