missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFBP1 Monoclonal Antibody (OTI7D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808311S
This item is not returnable.
View return policy
Description
FGFBP1 Monoclonal antibody specifically detects FGFBP1 in Human samples. It is validated for Western BlotSpecifications
| FGFBP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P31513 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2328 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7D10 | |
| Unconjugated | |
| Fmo3 | |
| 17 kDa HBGF-binding protein; 17 kDa heparin-binding growth factor-binding protein; FGF-binding protein 1; FGFBP; FGF-BP; Fgfbp1; FGF-BP1; FGFBP-1; fibroblast growth factor binding protein 1; fibroblast growth factor-binding protein 1; growth factor binding protein-1; growth factor-binding protein 1; HBP17; heparin-binding growth factor binding protein | |
| Human recombinant protein fragment corresponding to amino acids 1-291 of human FMO3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction