missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGFR1OP Recombinant Rabbit Monoclonal Antibody (JE64-24)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544761
This item is not returnable.
View return policy
Description
FGFR1OP Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
This gene encodes a largely hydrophilic protein postulated to be a leucine-rich protein family member. A t(6;8)(q27;p11) chromosomal translocation, fusing this gene and the fibroblast growth factor receptor 1 (FGFR1) gene, has been found in cases of myeloproliferative disorder. The resulting chimeric protein contains the N-terminal leucine-rich region of this encoded protein fused to the catalytic domain of FGFR1. This gene is thought to play an important role in normal proliferation and differentiation of the erythroid lineage. Alternatively spliced transcript variants that encode different proteins have been identified.
Specifications
| FGFR1OP | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O95684, Q4V7C1, Q66JX5 | |
| FGFR1OP | |
| Synthetic peptide within human FGFR1 Oncogene Partner aa 350-399/399. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JE64-24 | |
| Unconjugated | |
| FGFR1OP | |
| 4930553O10Rik; centrosomal protein 43; centrosomal protein 43; FGFR1 oncogene partner; CEP43; FGFR1 oncogene partner; FGFR1 oncogene partner 1; FGFR1 oncoprotein partner-like protein; Fgfr1op; fibroblast growth factor receptor 1 oncogene partner; FOP; LOC683722; Unknown (protein for MGC:134294) | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11116, 683722, 75296 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction