missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR2 Antibody (2E9), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | FGFR2 |
|---|---|
| Clone | 2E9 |
| Dilution | Western Blot 1:500-1:5000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043955
|
Novus Biologicals
NBP3-26583-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043495
|
Novus Biologicals
NBP3-26583-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGFR2 Monoclonal antibody specifically detects FGFR2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
| FGFR2 | |
| Western Blot 1:500-1:5000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25 | |
| A synthesized peptide derived from Human FGFR2 [UniProt P21802] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 2E9 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Protein Kinase | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2263 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title