missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR3 Antibody (1E3F9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52469
This item is not returnable.
View return policy
Description
FGFR3 Monoclonal specifically detects FGFR3 in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.
Specifications
| FGFR3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready | |
| EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism | |
| Mouse | |
| 87.7 kDa | |
| 0.1 mg | |
| Angiogenesis, Cancer, Protein Kinase | |
| 2261 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry | |
| 1E3F9 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| FGFR3 | |
| Purified recombinant fragment of human FGFR3 (AA: 529-694) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction