missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR3 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41227H
This item is not returnable.
View return policy
Description
FGFR3 Polyclonal antibody specifically detects FGFR3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| FGFR3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism | |
| Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human FGFR3. The immunogen is located within amino acids 630 - 680 of FGFR3. Amino Acid Squence: RDVHNLDYYKKTTNGRLPV | |
| 0.1 mL | |
| Angiogenesis, Cancer, Protein Kinase | |
| 2261 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction