Learn More
FGFR3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55358-1-AP-150UL
 View more versions of this product
Product Code. 18019815
  • € 410.00 / 150µL
Description
The FGFR3 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human FGFR3. This antibody recognizes human, mouse, rat antigen. The FGFR3 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
FGFR3
650 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Immunohistochemistry, ELISA
ACH, CD333, CEK2, FGFR 3, FGFR3, HSFGFR3EX, JTK4
150 μL
2261
Store at -20°C. Stable for one year after shipment.