missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR4 Antibody (SR2266), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21745-50ul
This item is not returnable.
View return policy
Description
FGFR4 Monoclonal antibody specifically detects FGFR4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| FGFR4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR2266 | |
| Western Blot 1:500-1:2000 | |
| CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase | |
| A synthesized peptide derived from human FGFR4 (Uniprot #: P22455) | |
| 50 μg | |
| Angiogenesis, Protein Kinase | |
| 2264 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction