missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR4 Antibody (U3-1784) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28842JF525
This item is not returnable.
View return policy
Description
FGFR4 Monoclonal antibody specifically detects FGFR4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| FGFR4 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase | |
| FGFR4/ CD334 | |
| 0.1 mL | |
| Angiogenesis, Protein Kinase | |
| 2264 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| U3-1784 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction