Learn More
FGFR4-IN-1, MedChemExpress
Product Code. 16473108
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16473108
Brand: Medchem Express HY100631
MedChemExpress FGFR4-IN-1 is a potent inhibiotr of FGFR4 with IC50 of 0.7 nM.
FGFR4-IN-1 is a potent inhibiotr of FGFR4 with IC50 of 0.7 nM.
Specifications
Chemical Name or Material FGFR4-IN-1
CAS 1708971-72-5
Color White
Purity Grade Notes Research
Molecular Formula C24H27N7O5
Quantity 10 mg
Solubility Information DMSO : 6.4 mg/mL (12.97 mM; Need warming)
SMILES O=C(N1CCCC2=C1N=C(C=O)C(CN3C(COCC3)=O)=C2)NC4=NC=C(C#N)C(NCCOC)=C4
Molecular Weight (g/mol) 493.52
Formula Weight 493.52
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only