missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR4 Rabbit anti-Macaque, Clone: 148, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90949-50UL
This item is not returnable.
View return policy
Description
FGFR4 Monoclonal specifically detects FGFR4 in Rhesus Macaque samples. It is validated for ELISA.
Specifications
| FGFR4 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Macaque | |
| Purified |
| ELISA | |
| 148 | |
| ELISA 1:5000-1:10000 | |
| CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase | |
| Recombinant Rhesus FGFR4 Protein | |
| 50 μg | |
| Angiogenesis, Protein Kinase | |
| 2264 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction