Learn More
FGG Monoclonal Antibody (OTI4G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA507154
Encompass_Preferred
 View more versions of this product
Product Code. 17516962
  • € 540.87 / 100µL
Description
The protein encoded by this gene is the gamma component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including dysfibrinogenemia, hypofibrinogenemia and thrombophilia. Alternative splicing results in two transcript variants encoding different isoforms.
Specifications
Specifications
FGG
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P02679
FGG
Full length human recombit protein of human FGG produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI4G2
Unconjugated
FGG
fibrinogen gamma chain
Mouse
Affinity Chromatography
RUO
2266
-20° C, Avoid Freeze/Thaw Cycles