Learn More
Invitrogen™ FGL2 Monoclonal Antibody (532223)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA561610
Product Code. 30535007
  • € 469.00 / 100µg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Fibrinogen-like protein 2 (Fgl2), also known as fibroleukin, is secreted by T cells and is involved in diseases in which thrombosis plays a pivotal role, such as virus-induced fulminant hepatitis, fetal loss syndrome and xenograft rejection. Constitutively expressed in cytotoxic T cells, Fgl2 exerts immunosuppressive effects on both T cell proliferation and dendritic cell maturation. Fgl2 is a serine protease and directly cleaves prothrombin to thrombin. Fgl2 functions in the pathogenesis of diseases including viral-induced hepatitis and Th1 cytokine-induced fetal loss syndrome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FGL2
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q14314
FGL2
Chinese hamster ovary cell line CHO-derived recombinant human FGL2, Asn24-Pro439.
100 μg
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
532223
Unconjugated
FGL2
AI385601; associated with MHV-3 induced prothrombinase activity; cytotoxic T-lymphocyte-specific protein; Fgl2; Fiblp; fibrinogen like 2; fibrinogen-like 2; fibrinogen-like protein 2; Fibroleukin; musfiblp; Prothrombinase; pT49; similar to Fibroleukin precursor (Fibrinogen-like protein 2) (pT49); T49
Mouse
Protein A/G
RUO
10875
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized