missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FGL2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17257993 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17257993 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17257993 Supplier Invitrogen™ Supplier No. PA5115230

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Fgl2.

Fibrinogen-like protein 2 (Fgl2), also known as fibroleukin, is secreted by T cells and is involved in diseases in which thrombosis plays a pivotal role, such as virus-induced fulminant hepatitis, fetal loss syndrome and xenograft rejection. Constitutively expressed in cytotoxic T cells, Fgl2 exerts immunosuppressive effects on both T cell proliferation and dendritic cell maturation. Fgl2 is a serine protease and directly cleaves prothrombin to thrombin. Fgl2 functions in the pathogenesis of diseases including viral-induced hepatitis and Th1 cytokine-induced fetal loss syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGL2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene FGL2
Gene Accession No. Q14314
Gene Alias AI385601; associated with MHV-3 induced prothrombinase activity; cytotoxic T-lymphocyte-specific protein; Fgl2; Fiblp; fibrinogen like 2; fibrinogen-like 2; fibrinogen-like protein 2; Fibroleukin; musfiblp; Prothrombinase; pT49; similar to Fibroleukin precursor (Fibrinogen-like protein 2) (pT49); T49
Gene Symbols FGL2
Host Species Rabbit
Immunogen A synthesized peptide derived from human FGL2(Accession Q14314), corresponding to amino acid residues L343-Y393.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10875, 84586
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.