missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHIT Monoclonal Antibody (OTI2C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811502
This item is not returnable.
View return policy
Description
Alterations and deletions of the FHIT gene are strongly linked to the genesis and establishment of human tumors of the lung, cervix, breast, colon, stomach and pancreas. In normal cells, FHIT may act as a tumor suppressor.Specifications
| FHIT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P49789 | |
| FHIT | |
| Full length human recombit protein of human FHIT produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2C1 | |
| Unconjugated | |
| FHIT | |
| AP3Aase, FRA3B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2272 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction