missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHIT Monoclonal Antibody (OTI2C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811502S
This item is not returnable.
View return policy
Description
FHIT Monoclonal antibody specifically detects FHIT in Human samples. It is validated for Western BlotSpecifications
| FHIT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P37058 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3293 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2C1 | |
| Unconjugated | |
| HSD17B3 | |
| AP3A hydrolase; AP3Aase; AW045638; bis(5'-adenosyl)-triphosphatase; bis5'-adenosyl-triphosphatase; Diadenosine 5',5'''-P1,P3-triphosphate hydrolase; dinucleosidetriphosphatase; FHIT; Fra14A2; FRA3B; fragile histidine triad; fragile histidine triad gene; fragile histidine triad protein | |
| Human recombinant protein fragment corresponding to amino acids 49-268 of human HSD17B3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction