missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHIT Rabbit anti-Human, Clone: 104, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90283-100UL
This item is not returnable.
View return policy
Description
FHIT Monoclonal specifically detects FHIT in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| FHIT | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| 104 | |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
| AP3A hydrolase, AP3AaseFRA3Bbis(5′-adenosyl)-triphosphatase, Diadenosine 5′-5′′′-P1, dinucleosidetriphosphatase, EC 3.6.1.29, fragile histidine triad gene, Fragile histidine triad protein, P3-triphosphate hydrolase, tumor suppressor protein | |
| Recombinant Human FHIT Protein | |
| 100 μg | |
| Apoptosis, Cancer, Cellular Markers, Lipid and Metabolism, Tumor Suppressors | |
| 2272 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction