missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHL1 Antibody (OTI3G7), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70739MFV610
This item is not returnable.
View return policy
Description
FHL1 Monoclonal antibody specifically detects FHL1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| FHL1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI3G7 | |
| Flow Cytometry | |
| bA535K18.1, four and a half LIM domains 1, KYOT, KYO-T, LIM protein SLIMMER, skeletal muscle LIM-protein1, SLIM1Isl-1 and Mec-3 domains 1 | |
| Full length human recombinant protein of human FHL1 (NP_001440) produced in HEK293T cell. | |
| 0.1 mL | |
| Apoptosis, Cancer, Lipid and Metabolism, Tumor Suppressors | |
| 2273 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction