missing translation for 'onlineSavingsMsg'
Learn More

FHL1 Monoclonal Antibody (OTI1D6), TrueMAB™, OriGene

Product Code. 17543841
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17543841 30 μL 30µL
17523841 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17543841 Supplier Origene Technologies Supplier No. TA501295S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FHL1 Monoclonal antibody specifically detects FHL1 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen FHL1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1D6
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CYB5R3
Gene Accession No. P00387
Gene Alias bA535K18.1; cb1039; fhl; Fhl1; FHL-1; FHL1A; fhl1b; fhla; FLH1A; four and a half LIM domains; four and a half LIM domains 1; four and a half LIM domains 1a; four and a half LIM domains 1b; four and a half LIM domains 1b; four and a half LIM domains protein 1b; four and a half LIM domains a; four and a half LIM domains protein 1; four and a half LIM domains protein 1; four and a half LIM domains protein 1a; four-and-a-half Lin11, Isl-1 and Mec-3 domains 1; KYOT; KYO-T; LIM protein; LIM protein SLIMMER; MGC111107; RAM14-1; RBMX1A; RBMX1B; RBP-associated molecule 14-1; RP11-535K18.1; skeletal muscle LIM-protein 1; SLIM; SLIM1; SLIM-1; SLIMMER; XMPMA; zgc:92025
Host Species Mouse
Immunogen Full length human recombinant protein of human CYV5R3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1727
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.